Learn More
R&D Systems™ CometAssay Silver Staining Kit

Description
- Using the CometAssay™ Silver Staining Component Kit, permanent records that can be visualized using standard light microscopy are prepared, thereby avoiding the problems associated with fluorescent stains and epifluorescence microscopy
- The CometAssay™ Silver Staining Components Kit were designed specifically for use with the CometAssay™ system and CometSlides™ to minimize unwanted background and the amount of hazardous waste generated by silver nitrate
Specifications
Specifications
| Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
| Format | Kit |
| Includes | Only includes silver staining components for use after performing the comet assay |
| No. of Samples | 200 |
| Product Type | Commet Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.