Learn More
R&D Systems™ CometAssay Single Cell Gel Electrophoresis Assay-Silver



Description
Includes
Lysis Solution, 2 x 500mL; Comet LMAgarose, 15mL; Trevigen CometSlide, 25/kit; pH 10 200mM EDTA, 12.5mL; Kit with Silver Stain (Mfr. No. 4251-050-K) also includes four staining reagents and a fixation additive.
The principle of the assay is based upon the ability of denatured, cleaved DNA fragments to migrate out of the nucleoid under the influence of an electric field, whereas undamaged DNA migrates slower and remains within the confines of the nucleoid when a
Choose the Neutral CometAssay to detect double-stranded breaks or the Alkaline CometAssay for a more sensitive protocol used to detect smaller amounts of damage including single and double-stranded breaks
Cells are immobilized in a bed of low melting poi
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 50 Tests |
| Product Type | Single Cell Gel Electrophoresis Assay |
| Content And Storage | Lysis Solution (2 x 500mL), Comet LMAgarose (15mL); Trevigen CometSlide (25/kit) and pH 10 200mM EDTA (12.5mL). The product requires storage at both 2 - 8° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Product Line | R&D Systems |
| Label or Dye | No Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.