Learn More
R&D Systems™ CometAssay Software
Description
- Quantitative Analysis using Comet™ Analysis Software
- Image capture
- Auto set the camera on untreated cells to avoid saturation of undamaged heads
- Position comet head to the left and comet tail to the right
- Take all images at same magnification (4X to 10X) and camera settings
- Save images as TIFF, JPEG or PNG
- Edit well images to remove comets not located within pattern of nonoverlapping micropores
- Determine median % DNA in the Tail for each of the 96 macrowells
- Untreated controls serve as a baseline for comparison to treated cells. Generally, <10% DNA in the tail is indicative of a healthy population
- Use boxplots to summarize data graphically
- Image capture
Specifications
Specifications
| Description | Simplifies assay performance from end-to-end, and assures sensitive, consistent quantitative results |
| For Use With (Equipment) | CometAssay™ System and high throughput CometChip™ technology, along with Microsoft Windows™ Operating System (Windows 10, Windows 8/8.1, Windows 7) |
| For Use With (Application) | Rapidly evaluate large numbers of cells, related to each treatment group or screening target in a study, and generate summary statistics based on the corresponding numeric results |
| Product Type | Analysis Software |