Description
- Includes:
- CometChip™ Starter Kit
- CometAssay™ Electrophoresis System II
Specifications
Specifications
| Content And Storage | The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Includes | CometAssay™ Electrophoresis System II, 96-Well CometChip™ System, CometChip™ 30 micron, 100 mL Lysis Solution, & 15 mL Comet LMAgarose |
| Product Line | R&D Systems |
| Product Type | ES Starter Kit+NA Power Supply |