Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ Series 130M Full Facepiece Respirators with Inner Mask
SDP

Catalog No. 19222476 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Includes:
Mask Only
Mask, Diskit™ P100 Cartridges
Mask, Diskit™ P100OV Cartridges
Mask, OV/95 Cartridges
Mask, OV/R95 Cartridges
Mask, P100 Cartridges
For Use With (Equipment):
Cartridges/Diskits
Dentec™ Filters and Cartridges
Filter Type:
N95
P100
R95
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes For Use With (Equipment) Filter Type
19-222-476 Mask, OV/95 Cartridges Cartridges/Diskits N95
19-222-475 Mask Only Dentec™ Filters and Cartridges -
19-222-477 Mask, OV/R95 Cartridges Cartridges/Diskits R95
19-222-478 Mask, P100 Cartridges Cartridges/Diskits P100
19-222-479 Mask, Diskit™ P100 Cartridges Cartridges/Diskits P100
19-222-480 Mask, Diskit™ P100OV Cartridges Cartridges/Diskits P100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-222-476 Supplier Dentec Safety™ Supplier No. 130M20DN5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Special compression silicone full facepiece respirators with inner mask feature large inhalation valves that allow for maximum inhalation. Available to purchase with or without cartridges and filter assemblies.

  • Silicone rubber
  • Large panoramic anti-scratch, anti-fog treated polycarbonate lens provides extended field of vision and is impact resistant
  • Quarter-sized inhalation valves allows the user to inhale a larger volume of air per breath, reducing breathing resistance
  • Metal speech diaphragm allows for communication
  • 5 point quick release buckles allows the user to quickly and easily don and doff the respirator
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator facepiece
  • Neckstrap allows user to hang product around the neck when needed
  • Optional prescription lens insert
  • Nylon storage and carry bag included

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Blue
Filter Type N95
Material Silicone
Product Type Pre-Assembled Full Face Respirator Kit
Size Medium
With Exhalation Valve Yes
Facepiece Material Silicone
For Use With (Equipment) Cartridges/Diskits
Includes Mask, OV/95 Cartridges
Lens Type Anti-fog
Model 130-M-20DN5
Suspension Type 5-point
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.