Learn More
Dentec Safety™ Comfort-Air™ Series 130M Full Facepiece Respirators with Inner Mask
Shop All Dentec Safety Specialists Inc. Products

Description
- Silicone rubber
- Large panoramic anti-scratch, anti-fog treated polycarbonate lens provides extended field of vision and is impact resistant
- Quarter-sized inhalation valves allows the user to inhale a larger volume of air per breath, reducing breathing resistance
- Metal speech diaphragm allows for communication
- 5 point quick release buckles allows the user to quickly and easily don and doff the respirator
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator facepiece
- Neckstrap allows user to hang product around the neck when needed
- Optional prescription lens insert
- Nylon storage and carry bag included
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Color | Blue |
| Filter Type | P100 |
| Material | Silicone |
| Product Type | Pre-Assembled Full Face Respirator Kit |
| Size | Medium |
| With Exhalation Valve | Yes |
| Facepiece Material | Silicone |
| For Use With (Equipment) | Cartridges/Diskits |
| Includes | Mask, P100 Cartridges |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.