Learn More
Dentec Safety™ Comfort-Air™Nx Filter Cover For N95 Filter Pads
SDP
Catalog No. 19222573 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Quantity:
350/Cs.
6 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-222-573 350/Cs.
19-222-572 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-573 Supplier Dentec Safety™ Supplier No. 14668BB350
Add to Cart
Add to Cart
Dentec Comfort-Air™Nx filter cover for N95 filter pads. Suitable for COVID virus in accordance with CDC guidelines and non-oil based aerosols. Can be mounted directly on all Comfort-Air™ series half and full face masks.
  • Ultra thin design will fit under any grinding shield or welding helmet
  • Will not obstruct field of vision
  • Allows user to conduct a positive and negative seal check
  • Protects N95 filters from damage
  • Extends filter life
  • Rugged and durable
  • Can be easily disinfected
Specifications
Disposable Reusable
Quantity 350/Cs.