Learn More
Dentec Safety™ Comfort-Air™Nx Filter Cover and Holder with N95 Filters
Shop All Dentec Safety Specialists Inc. Products
Description
- Ultra thin design will fit under any medical style or industrial grinding shield or welding helmet
- Will not obstruct field of vision
- Unique space plenums allows user to access the entire filter material reducing breathing resistance, providing longer cartridge life, reducing costs and improving productivity
- 'Saf-Twist' design quickly attaches the cartridge to all Dentec Safety half-mask or full face air-purifying respirator facepiece
- The offset connector improves visibility and allows face shields or welding helmets to be worn without interference
- Offset connector allows the user to rotate the cartridge into 3 different positions improves visibility and allows face shields or welding helmets to be worn without interference
- Fitler cover protects the filter extending filter life
Specifications
Specifications
| Color | Black |
| Disposable | Reusable |
| Filter Type | N95 |
| Material | Plastic |
| Product Type | N95 Filter with Cover and Holder |
| With Exhalation Valve | No |
| Latex Free | Yes |