Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech ComforTech™ Coveralls

Catalog No. 19811667 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-811-667 Large
19-811-665 Small
19-811-666 Medium
19-811-668 X-Large
19-811-669 2X-Large
19-811-670 3X-Large
19-811-671 4X-Large
19-811-672 5X-Large
19-811-673 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-811-667 Supplier AlphaProTech Supplier No. CVJ40223TH
This item is not returnable. View return policy

Coveralls constructed of lightweight, durable material that delivers protection against particles, biologicals, and fluids.

  • Lightweight
  • Durable
  • Low particle count
  • Serged seam coverall with thumb loops
  • Excellent filtration efficiency

Specifications

Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Finger Loops
Neck Style Collared
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
For Use With (Application) Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.