Learn More
AlphaProTech Critical Cover™ ComforTech™ Frocks
BIOPHARMA
Catalog No. 19166110 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-166-110 Large
19-166-115 Medium
19-166-111 X-Large
19-166-112 2X-Large
19-166-114 4X-Large
19-166-113 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-166-110 Supplier AlphaProTech Supplier No. FKJ24913
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cleanroom frocks feature knit collar and wrists, pockets, and zipper closure.
  • Provide a 99.9% filtration efficiency rate
  • Protects wearers from light chemical splash, particles and biologicals
  • Keeps wearers cool and comfortable
  • Breathable and lightweight
  • Durable
  • Meets controlled environment needs
  • Cut and sewn to an appropriate length to ensure full coverage
  • Nylon zipper option adds another level of particle holdout and protection
Specifications
Size Large
Material Microporous Fabric
Color White
Sterility Non-sterile
Closure Type Zipper
Cuff Style Knit
Latex Free Yes
Sleeve Style Raglan Sleeve
Length (English) 12.8 in.
Seam Type Serged