Learn More
Specifications
Specifications
| Gene ID (Entrez) | 149951 |
| Species | Human |
| Concentration | 0.2 mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP2-42124. It Usually Blocks The Antibody Activity Completely by Incubating The Peptide with Equal Volume of Antibody for 30 Min (at 37°C.) |
| Gene Alias | C20orf92, chromosome 20 open reading frame 92, COMM domain containing 7, COMM domain-containing protein 7, dJ1085F17.3, FLJ14987, MGC33315 |
| Gene Symbol | COMMD7 |
| Quantity | 0.05 mg |
| Research Category | Cancer, Cell Cycle and Replication |
| Storage Buffer | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
