Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Common gamma Chain/IL-2 R gamma Antibody Pair [HRP]
SDP

Catalog No. NB010045 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010045 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010045 Supplier R&D Systems™ Supplier No. NBP27950315PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody Pair

specifically detects in Mouse samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 24 pg/mL
Assay Sensitivity 23.44-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Line Novus
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Common gamma Chain/IL-2 R gamma.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency
Gene ID (Entrez) 3561
Gene Symbol IL2RG
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.25 mg/mL rabbit anti-Common gamma Chain/IL-2 R gamma monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse Common gamma Chain/IL-2 R gamma monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 77 ng of recombinant mouse Common gamma Chain/IL-2 R gamma.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target Common gamma Chain/IL-2 R gamma
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1.5 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.