Learn More
Thermo Scientific™ Temperature Control Connectivity Accessories
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
$124.10 - $311.00
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
13-265-176
|
Thermo Scientific™
422000000004 |
Software | RS-232 Communications Interface |
Each for $311.00
|
|
||||
13-875-517
|
Thermo Scientific™
1600033 |
1.8 m Interface Cable | Cold/Heated Bath Circulators |
Each for $161.20
|
|
||||
13-265-100
|
Thermo Scientific™
1600035 |
5 m Interface Cable | Cold/Heated Bath Circulators |
Each for $124.10
|
|
||||
Description
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Connect your Thermo Scientific temperature control solution with your application.- Available for temperature control devices equipped with communications
- Designed to work with your Thermo Scientific recirculating chiller or bath equipped with communications
- Available in popular configurations that match your device’s connections and interface with your process
- Take the guess work out of finding parts to match your recirculating chiller or bath
- Provides remote control of Thermo Scientific units equipped with an RS-232 communications interface
- Choose upper and lower temperature alarm limits and monitor system status with an alarm
- Record your results on a user selectable graph
- Select from easy–to–use product icons
- Create graphs and charts
- Easily configure ramping functions over time
- View pop-up alarm windows which display the error condition
- View a virtual controller screen
- PC compatible only