Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COMP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595547 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595547 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595547 Supplier Invitrogen™ Supplier No. PA595547
  • $582.00
    $562.65 / Each of 1

    Save $19.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human Hacat whole cell, human CACO-2 whole cell, human 293T whole cell, rat cartilage tissue, mouse cartilage tissue. IHC: mouse knee-joint tissue, rat knee-joint tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

COMP is a noncollagenous extracellular matrix protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene COMP
Gene Accession No. P35444, P49747, Q9R0G6
Gene Alias Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5
Gene Symbols COMP
Host Species Rabbit
Immunogen E.coli-derived human COMP recombinant protein (Position: Q505-A757).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12845, 1311, 25304
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.