Learn More
Thermo Scientific™ Compact Digital Dry Bath/Block Heater
Description
Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.
- Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
- Designed for precise temperature control
- Fast heating rate
- Multiple combinations of different sizes of blocks allows for maximum tube size versatility during experimental procedure
- 12V low voltage power input
- Two sizes to meet your application volume demands
- Convenient and easy-to-read digital display
- Small footprint
- Lightweight design
- Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
- Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.
Applications:
- Sample preparation
- Enzyme preservation
- Enzyme-substrate reactions
- DNA amplification
Required Accessory:
- Unit must be ordered with block(s)
Warranty and Services
1 Year, Replacement
Specifications
Specifications
| Certifications/Compliance | CE, cTUVus, RoHS |
| Controller Type | Digital |
| Capacity (Metric) | 0.2 mL |
| Lighting | LED |
| Plug Type | CN, EU, UK |
| Temperature Range (Metric) | Ambient +2° to 100°C |
| Type | Dry Block Heater |
| Heating Rate | 20 min. average heating rate (30° to 100°C, ambient at 25°C) |
| Includes | Dry bath, CN, EU,and UK style plugs, block lifter, plastic lid |
| Model | Compact Dry Bath S |
| Show More |
Warranty: 1-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.