Learn More
Chemglass Life Sciences Stirrer, Magnetic, Five Position, Compact, Dura-Mag, 230V
SDP
Supplier:  Chemglass Life Sciences CLS410004E
Encompass_Preferred
Dura-Mag digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment.Dura-Mag stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation.Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance.Optional factory installed motion alarm available.Speed range: 25-400RPM. Please note: An Optional Flask Centering Template for Corning Disposable 500mL Spinner flasks is available.Installing the drop on template ensures the Corning Disposable Spinner flasks are centered directly over the magnet for optimal stirring.
Catalog No. 50-000-83268
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.