Learn More
Thermo Scientific COMPASS CENTRIFUGE SOFTWARE
Supplier:  Thermo Scientific 45910
Encompass_Preferred
Centrifuge Accessories; Compass Run Simulation software; Thermo Scientific; For use with WX ultracentrifuges; Windows-based; Protocols can be developed, simulated and exported to centrifuge; Each
Catalog No. 50-870-354
Only null left
Add to Cart
Request a Quote