Learn More
Invitrogen™ Complement C3a Monoclonal Antibody (2898)

Description
MA1-40158 detects C3/C3a from human samples. MA1-40158 has been successfully used in Western blot and ELISA applications. The MA1-40158 immunogen is the C-terminus of C3. Positive control: Human plasma; Negative control: C3-depleted serum.
Specifications
Specifications
| Antigen | Complement C3a |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2898 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | C3 |
| Gene Accession No. | P01024 |
| Gene Alias | Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.