Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C3d Monoclonal Antibody (053A-1149.3.1.4 (1003))
GREENER_CHOICE

Catalog No. MA183965 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183965 Supplier Invitrogen™ Supplier No. MA183965
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Mouse anti Human C3d antibody, clone 053A-1149.3.1.4 recognizes human complement component 3d (C3d), a approximately 33 kDa polypeptide fragment generated over the course of complement activation where complement 3 (C3) convertases cleave C3 to C3b, which is further degraded into iC3b and C3dg/C3d.

C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3d
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 053A-1149.3.1.4 (1003)
Concentration 1.17 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% sodium azide; pH 8.4
Gene C3
Gene Accession No. P01024
Gene Alias Acylation stimulating protein; Acylation stimulating protein cleavage product; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; Cd3; chain fragment 2; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3c alpha' Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3a; complement component C3b; complement component C3d; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3; Unknown (protein for MGC:134562)
Gene Symbols C3
Host Species Mouse
Immunogen Purified C3d from human plasma.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.