Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3dg fragment Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986197
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6197 Supplier Bioss Supplier No. BS4875R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.

Specifications

Antigen Complement C3dg fragment
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C3
Gene Accession No. P01024
Gene Alias Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; Cd3; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha′ chain; Complement C3c alpha′ chain fragment 1; Complement C3c alpha′ chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; prepro-C3
Gene Symbols C3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Complement C3dg fragment.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.