Learn More
Bioss Complement C4 / C4a Monoclonal
SDP
Supplier:  Bioss BSM61317R
Encompass_Preferred
Complement C4 / C4a Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-64678