Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C4d Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA185168
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA185168 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA185168 Supplier Invitrogen™ Supplier No. PA185168
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heat-mediated antigen retrieval using Tris/EDTA (pH 8.5) is recommended for the staining of paraffin sections. Rabbit anti Human C4d antibody is specific for human complement split product C4d.

C4 is a component of the classical complement pathway and is split by activated C1s to C4a and C4b. Together C2a and C4b forms the classical C3 convertase. The molecular mass of C4 is 204 kDa and it consists of 3 subunits: a (97 kDa), b (75 kDa) and g (33 kDa).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C4d
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.1% ProClin 950
Gene C4A
Gene Accession No. P0C0L4, P0C0L5
Gene Alias acidic C4; acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4; c4 complement; C4A; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4D; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4A (Rodgers blood group); complement component 4B (Chido blood group); CPAMD2; CPAMD3; DADB-112B14.11; FLJ60561; MGC164979; MHC class III region complement; RG; Rodgers form of C4
Gene Symbols C4A, C4B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1241-1256 of C4 conjugated to Keyhole limpet Haemocyanin.
Purification Method Protein G
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100293534, 720, 721
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.