Learn More
Invitrogen™ Complement C5 Polyclonal Antibody

Description
PA1-40289 detects C5 from mouse samples. PA1-40289 has been successfully used in Western blot applications. The PA1-40289 imunogen is mouse C5.
Specifications
Specifications
| Antigen | Complement C5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | Hc |
| Gene Accession No. | P06684 |
| Gene Alias | anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905 |
| Gene Symbols | Hc |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.