Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement C9 Monoclonal Antibody (aE11)
GREENER_CHOICE

Catalog No. PIMA533373 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533373 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533373 Supplier Invitrogen™ Supplier No. MA533373
  • $1,274.00
    $1,014.65 / Each of 1

    Save $259.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL). This antibody reacts with a C9 neoantigen of the terminal complement complex (TCC). Application Notes: Functional Assay - This antibody inhibits platelet activation by antiphospholipid antibody serum. IF - Cryosections of 10 μm were dried, fixed in acetone at 4°C for 10 min, dried and blocked for 20 min with PBSA. Positive control: Mucosa from patients with H. Pylori.

C5b-9 membrane attack complexes are assembled from five precursor molecules in the serum. Proteolytic cleavage of C5 by C5 convertase generates C5b which initiates assembly of the C5b-9 complex. The last step of C5b-9 complex formation involves polymerization of C9 which accompanies insertion of the complex into the cell membrane. During formation of C5b-8 and C9 polymerization, neoantigens are generated which are unique to the C5b-9 complex and are not present on any of the individual native complex components. The complement regulatory proteins CD59 and complement S-protein can both prevent C5b-9 insertion into the cell membrane. The formed SC5b-9 complex is unable to attach to cells and is cytolytically inactive. C5b-9 is involved in the progression of chronic proteinuric renal disease by mediating continuous tubulointerstitial damage. Early tubulointerstitial injury in the remnant kidney can be improved when C5b-9 complex forming is abrogated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C9
Applications ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Monoclonal
Clone aE11
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene C9
Gene Accession No. P02748, P48770
Gene Alias ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor
Gene Symbols C9
Host Species Mouse
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100034183, 100037951, 735
Target Species Human, Pig, Horse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.