Learn More
Invitrogen™ Complement Factor Ba Monoclonal Antibody (014III-33.2.4.3)
Shop All Thermo Scientific Products

Description
This antibody Mouse anti Human Complement Factor B (Ba Fragment) antibody, clone 014III-33.2.4.3 recognizes the 30 kDa Ba fragment of 90 kDa human complement factor B, present in blood serum. C3b associates with complement factor B, inducing conformational change.
Specifications
Specifications
| Antigen | Complement Factor Ba |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | 014III-33.2.4.3 |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | BBS with 0.1% sodium azide |
| Gene | CFB |
| Gene Accession No. | P00751 |
| Gene Alias | AHUS4; ARMD14; BF; B-factor, properdin; BFD; C3 proaccelerator; C3 proactivator; C3/C5 convertase; CFAB; CFB; CFBD; complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; FB; FBI12; GBG; Glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; H2-Bf; PBF2; Properdin factor B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.