Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor Bb Monoclonal Antibody (10-09)
GREENER_CHOICE

Catalog No. PIMA528085
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528085 Supplier Invitrogen™ Supplier No. MA528085
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes the Bb fragment of 90 kDa human complement factor B, present in blood serum. C3b associates with complement factor B, inducing conformational change which enables complement factor D to cleave the N-terminal of complement factor B, the Ba subunit, leaving the 63 kDa Bb subunit associated with C3b and forming C3 convertase. Subunit Bb is involved in the proliferation of preactivated B lymphocytes. Conversely, Ba inhibits lymphocyte proliferation.

CFB is involved in hemolytic uremic syndrome atypical 4 (AHUS4), an atypical form of hemolytic uremic syndrome. It is a complex genetic disease characterized by microangiopathic hemolytic anemia, thrombocytopenia, renal failure and absence of episodes of enterocolitis and diarrhea. In contrast to typical hemolytic uremic syndrome, atypical forms have a poorer prognosis, with higher death rates and frequent progression to end-stage renal disease. Disease susceptibility is associated with variations affecting the gene represented in this entry. Susceptibility to the development of atypical hemolytic uremic syndrome can be conferred by mutations in various components of or regulatory factors in the complement cascade system. Other genes may play a role in modifying the phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor Bb
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 10-09
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% sodium azide
Gene CFB
Gene Accession No. P00751
Gene Alias AHUS4; ARMD14; BF; B-factor, properdin; BFD; C3 proaccelerator; C3 proactivator; C3/C5 convertase; CFAB; CFB; CFBD; Complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; FB; FBI12; GBG; Glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; H2-Bf; PBF2; properdin factor B
Gene Symbols CFB
Host Species Mouse
Immunogen Human factor B.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 629
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.