Learn More
ACROBiosystems ACROBiosystems Human Complement Factor D / CFD Protein, Fc Tag
Supplier: ACROBiosystems CFDH5256100UG
Recombinant Protein;100UG;Human Complement Factor D, Fc Tag (CFD-H5256) is expressed from human 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession # P00746-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.8 kDa. The protein migrates as 53 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.