Learn More
Electron Microscopy Sciences Micro Tissue Grinder Kit, Complete Kit
SDP
Supplier:  Electron Microscopy Sciences 64800
Encompass_Preferred
The kit of is 7 autoclavable Wheaton 33 low extractable borosilicate micro Tissue Grinders includes 1 ml Tenbroeck, 0.5 ml, 0.1 ml, and 0.2 ml, 1 ml Dounce, 2 ml Potter-Elvehjem, and 1 ml tappered tissue grinders. The 0.5 ml micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures. This particular tissue grinder mortar can be safely joined with another threaded glass component if needed, using the Wheaton Connection. The Kit is packed in high-density polyethylene case with foam inserts.
Catalog No. 50-190-0363