Learn More
Fisherbrand™ Complete Plumbing Assemblies for Lab Workstations
Catalog No. 06000873 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Air Service Plumbing Assembly
Nitrogen Service Plumbing Assembly
Vacuum Service Plumbing Assembly
Mounting:
Left side
Right side
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Mounting
06-000-873 Vacuum Service Plumbing Assembly Right side
06-000-869 Air Service Plumbing Assembly Left side
06-000-870 Vacuum Service Plumbing Assembly Left side
06-000-871 Nitrogen Service Plumbing Assembly Left side
06-000-872 Air Service Plumbing Assembly Right side
06-000-874 Nitrogen Service Plumbing Assembly Right side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-000-873
Add to Cart
Add to Cart
Fisherbrand™ Complete Plumbing Assemblies are designed to be used with Lab Workstations.
  • Service turret is color coded based on service
  • Service turret is a 135 degree turret with one polished chrome valve with a brass floating cone and integral brass seat.
  • Service turrets provide metering control of laboratory gases at pressures up to 60 PSI.
  • System service hoses carry services from teh overhead service panel through the upright of the table and to the shank of the fixture. Hoses extend 48 in. above the workstation.
  • System service hoses are color coded reinforced PVC
  • Male Quick Connects has a double end shut off plug that have an internal valve that shuts off when uncoupled.
Specifications
Description Right Side, Vacuum Service Complete Plumbing Assembly
Color Code Yellow
Material PVC, Chrome
Type Vacuum Service Plumbing Assembly
Includes Male Quick Connect, System Hose and Service Turret
Mounting Right side