Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Complex IV Human Activity Microplate Assay Kit, 96 Tests

Catalog No. 502585031
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-5031 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-5031 Supplier Abcam Supplier No. AB10991096TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Colorimetric enzyme activity assay for human COX IV in cell and tissue extracts, cited in over 16 publications.

  • Assay Type: Enzyme activity
  • Detection Method: Colorimetric
  • Reactive Species: Human
  • Sample Types: Cell culture extracts, Tissue
  • Applications: Enzyme activity assay
  • Key Features: Cited in > 16 publications; compatible with microplate readers

Specifications

Target Molecule Complex IV, Human
No. of Reactions 96 Tests
Reactivity Human
For Use With (Equipment) Microplate Reader
Format Microplate Assay Kit
Detection Method Colorimetric
Cross Reactivity Human
Sample Type Cell Culture Extracts, Tissue
Quantity 96 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.