Learn More
Invitrogen™ COX5A Monoclonal Antibody (6E9B12D5)

Description
Positive control: Isolated mitochondria from Human heart, Bovine heart, Rat heart, Mouse heart, and HepG2, Human embryonic lung-derived fibroblasts (strain MRC5), HeLa cells.
Specifications
Specifications
| Antigen | COX5A |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6E9B12D5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | HEPES buffered saline with 0.02% sodium azide |
| Gene | COX5A |
| Gene Accession No. | P00426, P11240, P12787, P20674 |
| Gene Alias | AA959768; CcOX; COX; Cox5a; COX5A protein; COX-VA; cytochrome c oxidase polypeptide Va; cytochrome c oxidase polypeptide, mitochondrial; cytochrome c oxidase subunit 5A; cytochrome c oxidase subunit 5A, mitochondrial; cytochrome c oxidase subunit Va; cytochrome c oxidase, subunit Va; mitochondrial cytochrome c oxidase subunit 5a; mitochondrial cytochrome c oxidase subunit Va; VA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.