Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Compound Bar Set
SDPSCI_ED

Catalog No. S00658 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00658 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00658 Supplier United Scientific™ Supplier No. BMSET04
Add to Cart
Add to Cart

Compound Bar Set is a classic demonstration of thermal expansion.

  • Quick and easy tools for demonstrating thermal expansion.
  • There are four bar combinations: Brass/Aluminum, Brass/Iron, Copper/Iron, and Brass/ Copper.
  • Since each metal has a different coefficient of thermal expansion, they will create a unique curve when paired with a different metal.
  • Metal bars are stamped with the identification of each metal, Aluminum (Al), Iron (Fe), Copper (Cu).
  • The Brass bar is not stamped.
  • Includes activity guide.

Specifications

Grade 6 to 12
Format Set
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.