Description
Includes
Bimetal bar on wooden handle; Instructions
- A great tool for teaching students about thermostatic principles
- Thermal compound bar can be used for a dramatic demonstration of the thermostatic principle by showing the unequal expansion of two different metals
- Bar bends when heated due to its dual composition
- Bar consists of a bi-metal strip mounted in a plastic handle
- Instructions provided
Specifications
Specifications
| For Use With (Application) | Demonstrating the thermostatic principle |
| Dimensions (L x W x H) | 10 x 0.75 x 0.75 in. |