Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Compound Benzoin Tincture, USP, Spectrum™ Chemical
SDP

Catalog No. 18601824 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Poly Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-824 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-824 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BE15020LTBL

BE150

Spectrum™ Chemical Compound Benzoin Tincture, USP is a pungent solution used as an oral mucosal protectan. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS Max % 100%
Packaging Poly Drum
Quantity 20 L
UN Number UN1170
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.