Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Compression Cap
SDP

Catalog No. 31501972 Shop All Kemtech Products
Change view
Click to view available options
Thread Size (Metric):
8 mm
13 mm
20 mm
24 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
31-501-972 8 mm
31-501-973 13 mm
31-501-974 20 mm
31-501-975 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-501-972 Supplier Synthware™ Supplier No. RC005
Add to Cart
Add to Cart

Compression Cap Replacements with a hole for use with a thermometer/inlet adaptors or microscale reaction vials.

Specifications

Color Red
For Use With (Application) Organic Synthesis
Material Phenolic Resin
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) 8 mm
Product Type Compression Cap
For Use With (Equipment) For Hickman Still Head side port
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.