Learn More
Justrite™ Compression Fittings w/Ferrules for HPLC Poly Manifold
Catalog No. 19822312 Shop All Justrite Safety Group Products
Change view
Click to view available options
Diameter (Metric) Outer:
2 mm
2.5 mm
3 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
19-822-312 3 mm
19-822-310 2 mm
19-822-311 2.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-312 Supplier Justrite™ Supplier No. 28123
Add to Cart
Add to Cart
Bring old equipment back into service with replacement parts. The Justrite™ Compression Fittings w/Ferrules for HPLC Poly Manifold, Pkg/6 are accessories for HPLC functions.
  • Compression fittings, sold in sets of six, accommodate hard-wall tubing size of 3.0-mm OD
  • Fittings are approximately 25-mm (1 in.) long and made of ETFE (ethylene tetrafluoroethylene) -fluorine based plastic
  • Includes ferrules for compression fit to the waste line
Specifications
Color Red
Diameter (Metric) Outer 3 mm
Material ETFE