Learn More
Cayman Chemical Concanamycin A, 80890-47-7, 100 µg
SDP
Supplier:  Cayman Chemical 11050100
Encompass_Preferred
Blocks lysosomal acidification through selective inhibition of the V-type ATPase (EC50s = 2.1-2.3 μM); blocks cell surface expression of viral glycoproteins without affecting their synthesis and, at 0.8 μM, prevents entry of influenza virus into cells
Catalog No. NC1199988
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.