Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Concanamycin B, H+-ATPase Inhibitor, Solid, >98% Purity, 500 μg

Catalog No. 502705968
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5968 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5968 Supplier Abcam Supplier No. AB144228500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent and selective vacuolar type H+-ATPase inhibitor with antiviral and apoptotic activity.

  • Molecular Weight: 852.1 Da
  • Purity: >98%
  • Physical Form: Solid
  • Mechanism of Action: Potent, selective vacuolar type H+-ATPase inhibitor
  • Biological Activity: Antibiotic agent; induces EGF-dependent apoptosis; shows potent antiviral effects (IC50 = 6 nM, HSV-1 replication)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Target Molecule H+-ATPase
Formulation 100% Concanamycin B
Quantity 500 μg
For Use With (Application) H+-ATPase inhibition, Apoptosis induction, Antiviral research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.