Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Concanamycin C, H+-ATPase Inhibitor, Solid, 100 μg

Catalog No. 502705967
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5967 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5967 Supplier Abcam Supplier No. AB144229100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent and selective vacuolar-type H+-ATPase inhibitor that shows antiproliferative and antifungal effects.

  • Molecular Weight: 823.1 Da
  • Purity: >98%
  • Form: Solid
  • Description: Potent, selective vacuolar-type H+-ATPase inhibitor. Macrolide antibiotic agent and analog of Concanamycin A (ab144227). Shows antiproliferative and potent antifungal effects.
  • Storage: Store at -20°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Buffer 100% Concanamycin C
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule H+ ATPase
Formulation 100% Concanamycin C
Quantity 100 μg
For Use With (Application) H+-ATPase inhibition, antiproliferative research, antifungal research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.