Learn More
Abcam Concanamycin C, H+-ATPase Inhibitor, Solid, 25 μg
Description
- Molecular Weight: 823.1 Da
- Purity: >98%
- Action: Potent, selective vacuolar-type H+-ATPase inhibitor
- Classification: Macrolide antibiotic agent
- Analogue: Concanamycin A (ab144227) analog
- Biological Effects: Shows antiproliferative and potent antifungal effects
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | H+-ATPase |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% Concanamycin C |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Quantity | 25 μg |
| For Use With (Application) | H+-ATPase inhibition, antiproliferative research, antifungal research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.