Learn More
Grainger Concrete Chain Saw Bar 16 Bar L
Supplier:  Grainger 14H226
Encompass_Preferred
The ICS Concrete Chain Saw Bar is a 16-inch bar for cutting concrete and masonry. It is made of heat-treated alloy steel and has a .404 pitch. The bar fits ICS concrete saws that use a 3/8" drive sprocket nose. It has a standard mount pattern to fit most ICS concrete saws. The bar weighs 2.8 pounds and measures 16 x 1.25 x 0.5 inches. It is made in the USA. This durable bar can cut concrete, brick, block and stone.
Catalog No. 50-275-0673