Learn More
Synthware™ Reflux Condenser, Jacketed
GREENER_CHOICESDP
Catalog No. 31502099 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No:
14/10
Catalog No. Standard Taper Joint No
31-502-099 14/10
1 options
Catalog No. 31-502-099 Supplier Synthware™ Supplier No. C201080
Only null left
Add to Cart
Add to Cart
Jacketed reflux condenser has I.D. of inner tube large enough to accept a thermometer
  • Supplied with one holed Nylon™ compression cap and one Viton™ O-ring
Specifications
Material Glass
Standard Taper Joint No 14/10
Description Hose Connection: 8mm O.D.
Length (Metric) Jacket 80 mm