Learn More
Applied Biosystems™ Conditioning Reagent Dx
Description
This Conditioning Reagent Dx is designed for use withApplied Biosystems Dx genetic analyzers.
Conditioning Reagent Dx is available as a ready-to-use pouch and is used for priming the polymer pump, washing the polymer pump between polymer type changes, and during instrument shut down. The pouch has adequate volume for a one-time use. The top part of the pouch fitment is sealed with a plastic film, which should be removed prior to installation on to the instrument.
This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.
Order Info
Each
Specifications
Specifications
| Starting Material | DNA, RNA |
| Quantity | 1 Pk. |
| Format | Pouch |
| For Use With (Equipment) | 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer |
| Product Type | Conditioning Reagent |
| Shipping Condition | Wet Ice |
| Content And Storage | Store at 2 - 8°C. |
For In Vitro Diagnostic Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.