Learn More
Applied Biosystems™ Conditioning Reagent, for 3500/SeqStudio™ Flex
Description
Conditioning Reagent is offered here as ready-to-use pouch compatible with 3500 and SeqStudio Flex series genetic analyzers. It is designed to be used for priming the polymer pump, washing the polymer pump between polymer type changes, and during instrument shut down. The pouch has adequate volume for a one-time use. Incorporated into the label is a radio frequency identification (RFID) tag. The top part of the pouch fitment is sealed with a plastic film, which should be removed prior to installation on the instrument.
Order Info
Each
Specifications
Specifications
| Starting Material | DNA |
| Quantity | 1 unit |
| Format | Pouch |
| For Use With (Equipment) | 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers |
| Product Type | Conditioning Reagent |
| Shipping Condition | Wet Ice |
| Content And Storage | Store pouches at 2 to 8°C. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.