Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Conductivity of Solutions Apparatus
SDPSCI_ED

Catalog No. s13134 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S13134 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13134 Supplier United Scientific™ Supplier No. CSA001
Only null left
Add to Cart
Add to Cart

This 3V conductivity tester demonstrates the electrical properties of various liquids. Includes Activity Guide.

  • The item consists of a molded plastic lamp socket mounted on a base with two binding posts for connecting a wire cord.
  • It comes complete with a bulb and glass tumbler.
  • Includes activity guide.

Specifications

Grade 5+
For Use With (Application) Electrical properties of liquids experiments
Includes A molded plastic lamp socket mounted on a base with two binding posts for connecting a wire cord. Comes complete with a bulb and glass tumbler. Includes activity guide.
Class Size 1 Lab Group
Product Type Physics Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.