Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 10 μS/cm at 25°C (4.7 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. 223616 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
6 x 20 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2236-16 500 mL Natural Poly Bottle
22364 120 mL Natural Poly Bottle
2236-100P 6 x 20 mL Pack
2236-20 20 x 20 mL Pack
2236-32 1 L Natural Poly Bottle
2236-1 4 L Natural Poly Bottle
R223600010F 10 L Cubitainer
R223600020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 2236-16 Supplier Ricca Chemical Company Supplier No. 223616
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 10μS/cm (10μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 5.0E-4
CAS Max % 100.0, 5.0E-4
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 4.7 ppm as NaCl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.