Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 5000 μS/cm at 25°C (2620 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. 22451 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2245-1 4 L Natural Poly Bottle
2245-16 500 mL Natural Poly Bottle
2245-32 1 L Natural Poly Bottle
2245-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2245-1 Supplier Ricca Chemical Company Supplier No. 22451
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
Name Note 5 mS/cm (5000 μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
Concentration or Composition Notes 2620ppm as NaCl
CAS Min % 98.0, 0.26
CAS Max % 100.0, 0.27
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 4 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.