Learn More
Description
- It ensures accurate, reliable measurements essential for quality control, research, and environmental monitoring.
- The convenient size supports multiple calibrations and easy handling, helping maintain device accuracy through stable calibration.
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard 300,000 μS/cm at 25°C |
| Packaging | Bottle |
| CAS | 7732-18-5, 7447-40-7, 26628-22-8 |
| Conductivity | 300,000 μS/cm |
| Type | Conductivity Standard |
| Molecular Weight (g/mol) | Unknown |
| Quantity | 250 mL |
| Percent Purity | Unknown |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.