Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Conductivity Tester
SDPSCI_ED

Catalog No. S139480 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139480 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139480 Supplier United Scientific™ Supplier No. CNDT01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simple tool used to measure conductivity.

This handheld device provides a qualitative visual indicator of the conductivity of different solutions, acids or salts.

  • Two brass probes can be touched to a solid substance, or immersed in a liquid, to show conductivity on green LEDs that range from 1 to 10
  • Each probe is 53 mm long with a diameter of 3 mm. Overall length of the meter and probes together is 14 cm
  • Includes a 12 V battery and operating instructions

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.