Learn More
Abcam Conduritol B Epoxide, Beta-Glucosidase Inhibitor, >98% Purity, 25 mg
Description
- Molecular weight: 162.14 Da
- Purity: >98%
- Form: Solid
- Inhibitor type: Selective, irreversible β-glucosidase inhibitor
- Potency: IC50 values are 1 μM for β-glucosidase and 100 μM for α-glucosidase
- Biological activity: Upregulates CD1d expression by THP-1 cells, raises intracellular glucosylceramide levels, and induces α-synuclein aggregation in the CNS in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Beta-Glucosidase, Alpha-Glucosidase |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% Conduritol B Epoxide |
| Quantity | 25 mg |
| For Use With (Application) | β-glucosidase inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.